Antimony ≥99.9999% (metals basis), polycrystalline lumps, Puratronic® 2-8 mm (0.08-0.3 in)
Supplier: Thermo Scientific Chemicals
|
Warning
|
10598-09
10598-18
10598-90
CAAA10598-09EA
148.05
CAD
CAAA10598-09
CAAA10598-18
CAAA10598-90
Antimony ≥99.9999% (metals basis), polycrystalline lumps, Puratronic® 2-8 mm (0.08-0.3 in)
Antimony
MDL: MFCD00134030
Purity: 99.9999%
CAS No.: 7440-36-0
Odor: Odorless
MDL No.: MFCD00134030
Formula:
Sb Boiling Pt: 1637 °C (1013 hPa) Melting Pt: 630 °C Density: 6.684 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134030 CAS Number: 7440-36-0 EINECS: 231-146-5 UN: 2871 ADR: 6.1,III Merck Index: 12,00733 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...