Nickel 99+% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)

Supplier: Thermo Scientific Chemicals
Danger

10595-CH 10595-CJ 10595-RX
CAAA10595-CHEA 173.8 CAD
CAAA10595-CH CAAA10595-CJ CAAA10595-RX
Nickel 99+% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)
Nickel

MDL: MFCD00011137


Purity: 99+% CAS No.: 7440-02-0 Physical Form: ≈67.8g/20x30cm, Temper: soft Odor: Odorless MDL No.: MFCD00011137

Formula: Ni
Boiling Pt: 3007 °C (1013 hPa)
Melting Pt: 1453 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR