Zirconium ≥99.5% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)
Supplier: Thermo Scientific Chemicals
10594-GM
10594-GR
10594-IB
CAAA10594-GMEA
167.48
CAD
CAAA10594-GM
CAAA10594-GR
CAAA10594-IB
Zirconium ≥99.5% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)
Zirconium
MDL: MFCD00011303
Purity: 99.5%
CAS No.: 7440-67-7
Physical Form: ≈10.3g/100x125mm, Temper: soft
Odor: Odorless
MDL No.: MFCD00011303
Formula:
Zr Storage Temperature: Ambient |
MDL Number:
MFCD00011303 CAS Number: 7440-67-7 EINECS: 231-176-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...