Zirconium ≥99.8% (metals basis excluding Hf), foil (Hf ∼ 2%), annealed, Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

10591-GI 10591-GR 10591-IB
CAAA10591-GIEA 103.17 CAD
CAAA10591-GI CAAA10591-GR CAAA10591-IB
Zirconium ≥99.8% (metals basis excluding Hf), foil (Hf ∼ 2%), annealed, Thickness: 0.25 mm (0.01 in)
Zirconium

MDL: MFCD00011303


Purity: 99.8% CAS No.: 7440-67-7 Physical Form: ≈20.6g/100x125mm, Temper: soft Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR