Zirconium ≥99.2% (metals basis excluding Hf), foil (max. 4.5% Hf), annealed, Thickness: 1.0 mm (0.039 in)

Supplier: Thermo Scientific Chemicals

10590-FI 10590-GH 10590-HP
CAAA10590-FIEA 94.33 CAD
CAAA10590-FI CAAA10590-GH CAAA10590-HP
Zirconium ≥99.2% (metals basis excluding Hf), foil (max. 4.5% Hf), annealed, Thickness: 1.0 mm (0.039 in)
Zirconium

MDL: MFCD00011303


Purity: 99.2% CAS No.: 7440-67-7 Physical Form: ≈16.3g/50x50mm, Temper: soft Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR