Zirconium ≥99.2% (metals basis excluding Hf), wire (max. 4.5% Hf), Ø 2.0 mm (0.08 in)

Supplier: Thermo Scientific Chemicals

10588-G1 10588-G5 10588-G9
CAAA10588-G1EA 111.71 CAD
CAAA10588-G1 CAAA10588-G5 CAAA10588-G9
Zirconium ≥99.2% (metals basis excluding Hf), wire (max. 4.5% Hf), Ø 2.0 mm (0.08 in)
Zirconium

MDL: MFCD00011303


Purity: 99.2% CAS No.: 7440-67-7 Physical Form: ≈20.4g/m Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR