Titanium ≥99.95% (metals basis), sponge 3-19 mm (0.12-0.75 in)
Supplier: Thermo Scientific Chemicals
|
Warning
|
10583-18
10583-30
10583-A1
10583-A7
CAAA10583-18EA
67.94
CAD
CAAA10583-18
CAAA10583-30
CAAA10583-A1
CAAA10583-A7
Titanium ≥99.95% (metals basis), sponge 3-19 mm (0.12-0.75 in)
Titanium
MDL: MFCD00011264
Purity: 99.95%
CAS No.: 7440-32-6
Appearance: Grey
Odor: Odorless
UN No.: 2878
MDL No.: MFCD00011264
Formula:
Ti MW: 47.88 g/mol Boiling Pt: 3262 Melting Pt: 1660 Density: 4.51 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 UN: 2878 ADR: 4.1 III Merck Index: 12,09610 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...