Titanium ≥99.7% (metals basis), sponge ≤ 25.4 mm (1.0 in)

Supplier: Thermo Scientific Chemicals
Warning

10582-18 10582-A1
CAAA10582-06EA 47.38 CAD
CAAA10582-06 CAAA10582-18 CAAA10582-A1
Titanium ≥99.7% (metals basis), sponge ≤ 25.4 mm (1.0 in)
Titanium

MDL: MFCD00011264


Purity: 99.7% CAS No.: 7440-32-6 Odor: Odorless UN No.: 2878 MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
UN: 2878
ADR: 4.1,III
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR