Nickel ≥99.8% (metals basis), spherical powder -300 mesh

Supplier: Thermo Scientific Chemicals
Danger

10581-09 10581-22 10581-36 10581-A3
CAAA10581-09EA 39.5 CAD
CAAA10581-09 CAAA10581-22 CAAA10581-36 CAAA10581-A3
Nickel ≥99.8% (metals basis), spherical powder -300 mesh
Nickel

MDL: MFCD00011137


Purity: 99.8% CAS No.: 7440-02-0 Odor: Odorless UN No.: 3089 MDL No.: MFCD00011137

Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR