Aluminium ≥99.999% (metals basis), ingot

Supplier: Thermo Scientific Chemicals

10571-22 10571-36 10571-A3
CAAA10571-22EA 140.94 CAD
CAAA10571-22 CAAA10571-36 CAAA10571-A3
Aluminium ≥99.999% (metals basis), ingot
Aluminium

MDL: MFCD00134029


Purity: 99.999% CAS No.: 7429-90-5 Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR