Ruthenium(III) acetylacetonate Ru ≥ 24%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ruthenium (III) 2,4-pentanedionate, Tris(2,4-pentanedionato-O,O')ruthenium (III), Ruthenium (III) tris(2,4-pentanedionato-O,O'), Tris(2,4-pentanedionato)ruthenium(III)
10568-03
10568-06
CAAA10568-03EA
172.22
CAD
CAAA10568-03
CAAA10568-06
Ruthenium(III) acetylacetonate Ru ≥ 24%
Ruthenium (III) acetylacetonate
Hydrogenation
CAS No.: 14284-93-6
Molecular Formula: C15H21O6Ru
Formula Weight: 398.40
Physical Form: Crystalline
Appearance: Red
Melting Point: 260° dec.
MDL No.: MFCD00000030
Hydrogenation
Formula:
C₁₅H₂₁O₆Ru MW: 398.4 g/mol Melting Pt: 260 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00000030 CAS Number: 14284-93-6 EINECS: 238-193-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...