Rubidium sulfate ≥99.8% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

10567-09 10567-18
CAAA10567-09EA 86.43 CAD
CAAA10567-09 CAAA10567-18
Rubidium sulfate ≥99.8% (metals basis)
Rubidium sulphate

MDL: MFCD00011190


Purity: 99.8% CAS No.: 7488-54-2 Molecular Formula: Rb2SO4 Formula Weight: 267.00 Physical Form: -20 Mesh Powder Density: 3.613 Refractive Index: 1.513 Odor: Odorless Melting Point: 1050° MDL No.: MFCD00011190

Formula: O₄Rb₂S
MW: 267 g/mol
Boiling Pt: 1708 °C (1013 hPa)
Melting Pt: 1050 °C
Density: 3,613
Storage Temperature: Ambient
MDL Number: MFCD00011190
CAS Number: 7488-54-2
EINECS: 231-301-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR