Rubidium hydroxide 50% (w/w) in aqueous solution , 99.6% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

10565-NC 10565-NE
CAAA10565-NCEA 232.26 CAD
CAAA10565-NC CAAA10565-NE
Rubidium hydroxide 50% (w/w) in aqueous solution , 99.6% (metals basis)
Rubidium hydroxide

MDL: MFCD00011194


Purity: 99.6+% CAS No.: 1310-82-3 Molecular Formula: HORb Formula Weight: 102.48(anhy) Physical Form: Liquid Appearance: Pale yellow Merck Reference: 14,8285 UN No.: 2677 MDL No.: MFCD00011194

Formula: RbOH
MW: 102.48 g/mol
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 300…302 °C
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011194
CAS Number: 1310-82-3
EINECS: 215-186-0
UN: 2677
ADR: 8,II
Merck Index: 14,8285

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR