Rubidium chloride ≥99.8% (metals basis)
Supplier: Thermo Scientific Chemicals
Warning
|
10563-09
10563-18
CAAA10563-09EA
135.56
CAD
CAAA10563-09
CAAA10563-18
Rubidium chloride ≥99.8% (metals basis)
Rubidium chloride
MDL: MFCD00011187
Purity: 99.8%
CAS No.: 7791-11-9
Molecular Formula: ClRb
Formula Weight: 120.92
Physical Form: -20 Mesh Crystalline
Density: 2.800
Refractive Index: 1.493
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 718°
Boiling Point: 1390°
Merck Reference: 14,8284
MDL No.: MFCD00011187
Formula:
RbCl Boiling Pt: 1390 °C Melting Pt: 718 °C Density: 2.800 Storage Temperature: Ambient |
MDL Number:
MFCD00011187 CAS Number: 7791-11-9 EINECS: 232-240-9 Merck Index: 14,8284 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...