Aluminium ≥99.45% (metals basis), foil 30x30 cm (12x12 in), Thickness: 0.025 mm (0.001 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminum

10558-KJ 10558-KM 10558-KN
CAAA10558-KJEA 121.66 CAD
CAAA10558-KJ CAAA10558-KM CAAA10558-KN
Aluminium ≥99.45% (metals basis), foil 30x30 cm (12x12 in), Thickness: 0.025 mm (0.001 in)
Aluminium

MDL: MFCD00134029


Purity: 99.45% CAS No.: 7429-90-5 Physical Form: ≈6.17g/30x30cm Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR