Tetrakis(triphenylphosphine)platinum(0) Pt ≥ 15,2%

Supplier: Thermo Scientific Chemicals

Synonyms: Tetrakis(triphenylphosphine)platinum

10549-03 10549-06
CAAA10549-03EA 237 CAD
CAAA10549-03 CAAA10549-06
Tetrakis(triphenylphosphine)platinum(0) Pt ≥ 15,2%
Tetrakis(triphenylphosphine)platinum (0)

Hydrosilation and oxidation


CAS No.: 14221-02-4 Molecular Formula: Pt[P(C6H5)3]4 Formula Weight: 1244.21 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Sensitivity: Air Sensitive Appearance: Yellow Odor: Odorless Solubility: Soluble in benzene MDL No.: MFCD00010014


Hydrosilation and oxidation

Formula: C₇₂H₆₀P₄Pt
MW: 1244.24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00010014
CAS Number: 14221-02-4
EINECS: 238-087-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR