Potassium tetranitroplatinate(II) Pt ≥ 42,6%

Supplier: Thermo Scientific Chemicals

10538-03 10538-06
CAAA10538-03EA 211.72 CAD
CAAA10538-03 CAAA10538-06
Potassium tetranitroplatinate(II) Pt ≥ 42,6%
Potassium tetranitroplatinate (II)

MDL: MFCD00011375


CAS No.: 13815-39-9 Molecular Formula: K2Pt(NO2)4 Formula Weight: 457.32 Physical Form: Crystalline MDL No.: MFCD00011375

Formula: K₂N₄O₈Pt
Storage Temperature: Ambient
MDL Number: MFCD00011375
CAS Number: 13815-39-9
EINECS: 237-491-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR