Phosphoryl trichloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Phosphorus (V) oxide chloride, Phosphorus oxychloride, Phosphoryl chloride, Phosphorus(V) oxychloride
10525-A1
10525-30
CAAA10525-A1EA
208.56
CAD
CAAA10525-A1
CAAA10525-30
Phosphoryl trichloride 99%
Phosphoryl trichloride
MDL: MFCD00011443
Purity: 99%
CAS No.: 10025-87-3
Molecular Formula: Cl3OP
Formula Weight: 153.33
Physical Form: Liquid
Density: 1.645
Refractive Index: 1.460
Sensitivity: Moisture Sensitive
Appearance: Colorless
Odor: Pungent
Melting Point: 1.25°
Boiling Point: 105.8°
Merck Reference: 14,7349
UN No.: 1810
MDL No.: MFCD00011443
Formula:
POCl₃ MW: 153.33 g/mol Boiling Pt: 105.8 °C Melting Pt: 1.25 °C Density: 1.645 Storage Temperature: Ambient |
MDL Number:
MFCD00011443 CAS Number: 10025-87-3 EINECS: 233-046-7 UN: 1810 ADR: 6.1,I Merck Index: 14,7349 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...