Diphosphorus pentoxide 98 ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phosphorus(V) oxide, di-Phosphorus pentoxide, Phosphorus pentoxide, Phosphoric anhydride

10524-18 10524-36
CAAA10524-18EA 70.47 CAD
CAAA10524-18 CAAA10524-36
Diphosphorus pentoxide 98 ACS
di-Phosphorus pentoxide

As a drying and dehydrating agent, as a condensing agent in organic synthesis, in sugar refining, as a laboratory reagent, and in fire extinguishing


Purity: 98% min CAS No.: 1314-56-3 Molecular Formula: P2O5 Formula Weight: 141.94 Physical Form: Powder Density: 2.39 Appearance: White Odor: Weak Melting Point: 340° Boiling Point: 360° subl. Solubility: Decomposes in water. Soluble in H2SO4. Insoluble in acetone and NH3 Merck Reference: 14,7355 UN No.: 1807 MDL No.: MFCD00011440


As a drying and dehydrating agent, as a condensing agent in organic synthesis, in sugar refining, as a laboratory reagent, and in fire extinguishing

Formula: P₂O₅
MW: 141.94 g/mol
Boiling Pt: 360 °C (subl.)
Melting Pt: 562
Density: 2.39
Storage Temperature: Ambient
MDL Number: MFCD00011440
CAS Number: 1314-56-3
EINECS: 215-236-1
UN: 1807
ADR: 8 II
Merck Index: 14,7355

Order Now

Specification Test Results

Insoluble matter 0.02%
P2O3 Pass test. (limit about 0.02%)
NH3 0.01%
Heavy Metals (as Pb) 0.01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR