Phosphorus(V) chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

10523-22 10523-36 10523-57
CAAA10523-22EA 79.95 CAD
CAAA10523-22 CAAA10523-36 CAAA10523-57
Phosphorus(V) chloride 98%
Phosphorus (V) chloride

As dehydrating and chlorinating agent, catalyst


Purity: 98% CAS No.: 10026-13-8 Molecular Formula: PCl5 Formula Weight: 208.24 Physical Form: Solid Density: 2.114 Appearance: Yellow Odor: Pungent Melting Point: 162° subl. Solubility: Soluble in carbon disulfide and carbon tetrachloride Merck Reference: 14,7351 UN No.: 1806 MDL No.: MFCD00011439


As dehydrating and chlorinating agent, catalyst

Formula: Cl₅P
Melting Pt: 162 °C (subl.)
Density: 2.114
Storage Temperature: Ambient
MDL Number: MFCD00011439
CAS Number: 10026-13-8
EINECS: 233-060-3
UN: 1806
ADR: 8,II
Merck Index: 14,7351

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR