Germanium(IV) oxide ≥99.9999% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Germanium dioxide

10510-06 10510-14 10510-22
CAAA10510-06EA 139.36 CAD
CAAA10510-06 CAAA10510-14 CAAA10510-22
Germanium(IV) oxide ≥99.9999% (metals basis)
Germanium (IV) oxide

MDL: MFCD00011030


Purity: 99.9999% CAS No.: 1310-53-8 Molecular Formula: GeO2 Formula Weight: 104.59 Physical Form: -40 Mesh Powder Density: 6.239 Appearance: White Odor: Odorless Melting Point: 1086° Merck Reference: 14,4409 MDL No.: MFCD00011030

Formula: GeO₂
MW: 104.61 g/mol
Melting Pt: 1086 °C
Density: 6.239
Storage Temperature: Ambient
MDL Number: MFCD00011030
CAS Number: 1310-53-8
EINECS: 215-180-8
Merck Index: 14,4409

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR