Germanium(IV) chloride ≥99.9999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
10509-06
10509-14
10509-22
CAAA10509-06EA
142.52
CAD
CAAA10509-06
CAAA10509-14
CAAA10509-22
Germanium(IV) chloride ≥99.9999% (metals basis)
Germanium (IV) chloride
MDL: MFCD00011029
Purity: 99.9999%
CAS No.: 10038-98-9
Molecular Formula: Cl4Ge
Formula Weight: 214.40
Physical Form: Liquid
Density: 1.874
Appearance: Colorless
Odor: Acidic
Melting Point: -49.5°
Boiling Point: 83.1°
Merck Reference: 14,4410
UN No.: 3264
MDL No.: MFCD00011029
Formula:
Cl₄Ge Boiling Pt: 83.1 °C Melting Pt: –49.5 °C Density: 1.874 Storage Temperature: Ambient |
MDL Number:
MFCD00011029 CAS Number: 10038-98-9 EINECS: 233-116-7 UN: 3264 ADR: 8,II Merck Index: 14,4410 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...