trans-Platin Pt ≥ 64,5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
10472-01
10472-03
10472-06
CAAA10472-01EA
107.44
CAD
CAAA10472-01
CAAA10472-03
CAAA10472-06
trans-Platin Pt ≥ 64,5%
trans-Platin
MDL: MFCD00011623
CAS No.: 14913-33-8
Molecular Formula: Cl2H6N2Pt
Formula Weight: 300.06
Physical Form: Crystalline
Appearance: Yellow
Odor: Odorless
MDL No.: MFCD00011623
Formula:
H₆Cl₂N₂Pt Melting Pt: 340 °C (dec.) Density: 3,746 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011623 CAS Number: 14913-33-8 EINECS: 238-980-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...