cis-Platin Pt ≥ 64.5% for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Platinol, cis-Diaminodichloroplatinum(II), cis-Diamminedichloroplatinum (II), cis-Dichlorodiammine platinum(II), Cisplatin, cis-Diamineplatinum(II) dichloride

10471-01 10471-03 10471-06
CAAA10471-01EA 243.32 CAD
CAAA10471-01 CAAA10471-03 CAAA10471-06
cis-Platin Pt ≥ 64.5% for NMR spectroscopy
cis-Platin

Potent anticancer agent that blocks DNA synthesis


CAS No.: 15663-27-1 Molecular Formula: Cl2H6N2Pt Formula Weight: 300.06 Physical Form: Micro Crystals Appearance: Yellow Odor: Odorless Merck Reference: 14,2317 UN No.: 3288 MDL No.: MFCD00011623


Potent anticancer agent that blocks DNA synthesis

Formula: H₆Cl₂N₂Pt
MW: 300.05 g/mol
Melting Pt: 270 °C (dec.)
Density: 3.738 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011623
CAS Number: 15663-27-1
EINECS: 239-733-8
UN: 3288
ADR: 6.1 II
Merck Index: 14,2317

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR