Aluminium oxide, γ-α-phase ≥99.98% (metals basis)
Supplier: Thermo Scientific Chemicals
10459-14
10459-22
10459-36
CAAA10459-14EA
93.54
CAD
CAAA10459-14
CAAA10459-22
CAAA10459-36
Aluminium oxide, γ-α-phase ≥99.98% (metals basis)
Aluminium oxide, γ-α-phase
MDL: MFCD00003424
Notes: Predominantly ? phase, 5-20% ? phase
Purity: 99.98%
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form: 0.01-0.02 Micron Powder, S.A. 100m2/g
Density: 3.965
Melting Point: 2045°
Boiling Point: 2980°
Merck Reference: 14,356
MDL No.: MFCD00003424
Formula:
Al₂O₃ MW: 101.96 Boiling Pt: 2980 Melting Pt: 2050 Density: 3.965 Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 Merck Index: 14,356 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...