Cobalt ≥99.5% (metals basis), powder -60 mesh

Supplier: Thermo Scientific Chemicals
Danger

10456-22 10456-36
CAAA10456-22EA 184.86 CAD
CAAA10456-22 CAAA10456-36
Cobalt ≥99.5% (metals basis), powder -60 mesh
Cobalt

MDL: MFCD00010935


Purity: 99.5% CAS No.: 7440-48-4 Appearance: Black Odor: Odorless UN No.: 3089 MDL No.: MFCD00010935

Formula: Co
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR