Cobalt ≥99.5% (metals basis), powder 1.6 µm

Supplier: Thermo Scientific Chemicals
Danger

10455-09 10455-22 10455-36
CAAA10455-09EA 133.51 CAD
CAAA10455-09 CAAA10455-22 CAAA10455-36
Cobalt ≥99.5% (metals basis), powder 1.6 µm
Cobalt

MDL: MFCD00010935


Purity: 99.8% CAS No.: 7440-48-4 Appearance: Black Odor: Odorless UN No.: 3089 MDL No.: MFCD00010935

Formula: Co
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR