Chromium ≥99.996% (metals basis), powder -10+20 mesh

Supplier: Thermo Scientific Chemicals

10452-06 10452-14 10452-22
CAAA10452-06EA 140.3 CAD
CAAA10452-06 CAAA10452-14 CAAA10452-22
Chromium ≥99.996% (metals basis), powder -10+20 mesh
Chromium

MDL: MFCD00010944


Purity: 99.996% CAS No.: 7440-47-3 Odor: Odorless MDL No.: MFCD00010944

Formula: Cr
MW: 52
Boiling Pt: 2469
Melting Pt: 1840
Density: 7.2
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
UN: 3089
ADR: 4.1 III
Merck Index: 12,02288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR