Zirconium ≥99% (metals basis excluding Hf), rods (max. 4.5% Hf), annealed, Ø 6.2 mm (0.24 in)

Supplier: Thermo Scientific Chemicals

10443-BS 10443-BW
CAAA10443-BSEA 97.96 CAD
CAAA10443-BS CAAA10443-BW
Zirconium ≥99% (metals basis excluding Hf), rods (max. 4.5% Hf), annealed, Ø 6.2 mm (0.24 in)
Zirconium

MDL: MFCD00011303


Purity: 99% CAS No.: 7440-67-7 Physical Form: ≈1.96g/cm, Temper: soft Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR