Zinc ≥99.9% (metals basis), foil, Width: 30 cm (12 in), Thickness: 0.62 mm (0.024 in)
Supplier: Thermo Scientific Chemicals
10437-BW
10437-CB
CAAA10437-BWEA
194.34
CAD
CAAA10437-BW
CAAA10437-CB
Zinc ≥99.9% (metals basis), foil, Width: 30 cm (12 in), Thickness: 0.62 mm (0.024 in)
Zinc
MDL: MFCD00011291
Purity: 99.9%
CAS No.: 7440-66-6
Physical Form: ≈665g/30x50cm
Odor: Odorless
MDL No.: MFCD00011291
Formula:
Zn Boiling Pt: 908 °C (1013 hPa) Melting Pt: 420 °C Density: 7.14 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...