Zinc ≥99.99% (metals basis), rods, Ø 1.27 cm (0.5 in)

Supplier: Thermo Scientific Chemicals

10433-BV 10433-RM 10433-RS
CAAA10433-BVEA 157.53 CAD
CAAA10433-BV CAAA10433-RM CAAA10433-RS
Zinc ≥99.99% (metals basis), rods, Ø 1.27 cm (0.5 in)
Zinc

MDL: MFCD00011291


Purity: 99.99+% CAS No.: 7440-66-6 Physical Form: ≈9.03g/cm Odor: Odorless MDL No.: MFCD00011291

Formula: Zn
Boiling Pt: 908 °C (1013 hPa)
Melting Pt: 420 °C
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR