Tungsten ≥99.95% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Supplier: Thermo Scientific Chemicals
10416-FI
10416-GH
10416-HW
CAAA10416-FIEA
127.19
CAD
CAAA10416-FI
CAAA10416-GH
CAAA10416-HW
Tungsten ≥99.95% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Tungsten
MDL: MFCD00011461
Purity: 99.95%
CAS No.: 7440-33-7
Physical Form: ≈4.8g/50x50mm
MDL No.: MFCD00011461
Formula:
W Boiling Pt: 5660 °C (1013 hPa) Melting Pt: 3410 °C Density: 19 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011461 CAS Number: 7440-33-7 EINECS: 231-143-9 Merck Index: 12,09945 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...