Tungsten ≥99.95% (metals basis), foil, Thickness: 0.5 mm (0.02 in)
Supplier: Thermo Scientific Chemicals
10414-FG
10414-FY
10414-GD
CAAA10414-FGEA
194.34
CAD
CAAA10414-FG
CAAA10414-FY
CAAA10414-GD
Tungsten ≥99.95% (metals basis), foil, Thickness: 0.5 mm (0.02 in)
Tungsten
MDL: MFCD00011461
Purity: 99.95%
CAS No.: 7440-33-7
Physical Form: ≈12g/25x50mm
MDL No.: MFCD00011461
Formula:
W MW: 183.84 g/mol Boiling Pt: 5660 Melting Pt: 3410 Density: 19 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011461 CAS Number: 7440-33-7 EINECS: 231-143-9 Merck Index: 12,09945 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...