Tungsten ≥99.95% (metals basis), wire, Ø 0.75 mm (0.03 in)

Supplier: Thermo Scientific Chemicals

10410-G1 10410-G5
CAAA10410-G1EA 99.7 CAD
CAAA10410-G1 CAAA10410-G5
Tungsten ≥99.95% (metals basis), wire, Ø 0.75 mm (0.03 in)
Tungsten

MDL: MFCD00011461


Purity: 99.95% CAS No.: 7440-33-7 Physical Form: ≈8.52g/m MDL No.: MFCD00011461

Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660
Melting Pt: 3410
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR