Tungsten ≥99.95% (metals basis), wire, Ø 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

10408-G6 10408-H2 10408-H7
CAAA10408-G6EA 98.43 CAD
CAAA10408-G6 CAAA10408-H2 CAAA10408-H7
Tungsten ≥99.95% (metals basis), wire, Ø 0.25 mm (0.01 in)
Tungsten

MDL: MFCD00011461


Purity: 99.95% CAS No.: 7440-33-7 Physical Form: ≈0.95g/m MDL No.: MFCD00011461

Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660
Melting Pt: 3410
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR