Tungsten ≥99.95% (metals basis), rods, Ø 12.7 mm (0.5 in)

Supplier: Thermo Scientific Chemicals

10403-BQ 10403-BU
CAAA10403-BQEA 281.24 CAD
CAAA10403-BQ CAAA10403-BU
Tungsten ≥99.95% (metals basis), rods, Ø 12.7 mm (0.5 in)
Tungsten

MDL: MFCD00011461


Purity: 99.95% CAS No.: 7440-33-7 Physical Form: ≈24.4g/cm MDL No.: MFCD00011461

Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660
Melting Pt: 3410
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR