Tungsten ≥99.9% (metals basis), powder APS 12 µm

Supplier: Thermo Scientific Chemicals
Warning

10401-09 10401-22 10401-36 10401-A3
CAAA10401-09EA 56.09 CAD
CAAA10401-09 CAAA10401-22 CAAA10401-36 CAAA10401-A3
Tungsten ≥99.9% (metals basis), powder APS 12 µm
Tungsten

MDL: MFCD00011461


Purity: 99.9% CAS No.: 7440-33-7 Physical Form: Powder Appearance: Grey UN No.: 3089 MDL No.: MFCD00011461

Formula: W
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR