Tungsten ≥99.9% (metals basis), powder APS 1-5 µm
Supplier: Thermo Scientific Chemicals
|
Warning
|
10400-09
10400-22
10400-36
10400-A3
CAAA10400-09EA
75.84
CAD
CAAA10400-09
CAAA10400-22
CAAA10400-36
CAAA10400-A3
Tungsten ≥99.9% (metals basis), powder APS 1-5 µm
Tungsten
MDL: MFCD00011461
Purity: 99.9%
CAS No.: 7440-33-7
Physical Form: Powder
Appearance: Grey
UN No.: 3089
MDL No.: MFCD00011461
Formula:
W MW: 183.84 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011461 CAS Number: 7440-33-7 EINECS: 231-143-9 UN: 3089 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...