Titanium ≥99.7% (metals basis), foil, annealed, Thickness: 0.89 mm (0.035 in)
Supplier: Thermo Scientific Chemicals
10399-GW
10399-ZB
10399-ZB
10399-GW
10399-GH
10399-GH
CAAA10399-ZBEA
659
CAD
CAAA10399-ZB
AA10399-GH
AA10399-GW
CAAA10399-GH
CAAA10399-GW
AA10399-ZB
Titanium ≥99.7% (metals basis), foil, annealed, Thickness: 0.89 mm (0.035 in)
Titanium
Formula:
Ti MW: 47.88 g/mol Boiling Pt: 3262 °C (1013 hPa) Melting Pt: 1660 °C Density: 4.51 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 Merck Index: 12,09610 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...