Titanium ≥99.2% (metals basis), foil, annealed, Thickness: 2.0 mm (0.08 in)

Supplier: Thermo Scientific Chemicals
Warning

10398-GH 10398-GW
CAAA10398-GHEA 128.14 CAD
CAAA10398-GH CAAA10398-GW
Titanium ≥99.2% (metals basis), foil, annealed, Thickness: 2.0 mm (0.08 in)
Titanium

MDL: MFCD00011264


Purity: 99.2% CAS No.: 7440-32-6 Physical Form: ≈90g/100x100mm Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR