Titanium 99,5 Metals basis, rods, annealed, Ø 12.7 mm (0.5 in)

Supplier: Thermo Scientific Chemicals
Warning

10394-BS 10394-BW 10394-CB
CAAA10394-BSEA 85.48 CAD
CAAA10394-BS CAAA10394-BW CAAA10394-CB
Titanium 99,5 Metals basis, rods, annealed, Ø 12.7 mm (0.5 in)
Titanium

MDL: MFCD00011264


Purity: 99.5% CAS No.: 7440-32-6 Physical Form: ≈5.7g/cm Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR