Titanium ≥99.7% (metals basis), rods, Ø 6.4 mm (0.25 in)

Supplier: Thermo Scientific Chemicals
Warning

10393-BU 10393-BY 10393-CQ
CAAA10393-BUEA 67.78 CAD
CAAA10393-BU CAAA10393-BY CAAA10393-CQ
Titanium ≥99.7% (metals basis), rods, Ø 6.4 mm (0.25 in)
Titanium

MDL: MFCD00011264


Purity: 99.7% CAS No.: 7440-32-6 Physical Form: ≈1.45g/cm Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR