Titanium ≥99.99% (metals basis), wire, annealed, Ø 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals
Warning

10389-BW 10389-CB 10389-CD
CAAA10389-BWEA 132.88 CAD
CAAA10389-BW CAAA10389-CB CAAA10389-CD
Titanium ≥99.99% (metals basis), wire, annealed, Ø 0.5 mm (0.02 in)
Titanium

MDL: MFCD00011264


Purity: 99.99% CAS No.: 7440-32-6 Physical Form: ≈0.88g/m, Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR