Titanium ≥99.5% (metals basis), foil, annealed, Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals
Warning

10385-GH 10385-HP 10385-HY
CAAA10385-GHEA 121.34 CAD
CAAA10385-GH CAAA10385-HP CAAA10385-HY
Titanium ≥99.5% (metals basis), foil, annealed, Thickness: 0.25 mm (0.01 in)
Titanium

MDL: MFCD00011264


Purity: 99.5% CAS No.: 7440-32-6 Physical Form: ≈11.3g/100x100mm Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR