Tin ≥99.8% (metals basis), shot 8-20 mesh
Supplier: Thermo Scientific Chemicals
10377-22
10377-36
10377-A3
CAAA10377-22EA
102.7
CAD
CAAA10377-22
CAAA10377-36
CAAA10377-A3
Tin ≥99.8% (metals basis), shot 8-20 mesh
Tin
MDL: MFCD00133862
Purity: 99.8%
CAS No.: 7440-31-5
Odor: Odorless
MDL No.: MFCD00133862
Formula:
Sn Boiling Pt: 2270 °C (1013 hPa) Melting Pt: 232 °C Density: 7.31 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 Merck Index: 12,09587 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...