Tellurium ≥99.999% (metals basis), lumps

Supplier: Thermo Scientific Chemicals
Warning

10362-14 10362-22 10362-36
CAAA10362-14EA 99.22 CAD
CAAA10362-14 CAAA10362-22 CAAA10362-36
Tellurium ≥99.999% (metals basis), lumps
Tellurium

MDL: MFCD00134062


Purity: 99.999+% CAS No.: 13494-80-9 Odor: Odorless MDL No.: MFCD00134062

Formula: Te
Boiling Pt: 990 °C (1013 hPa)
Melting Pt: 450 °C
Density: 8.83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
UN: 3284
ADR: 6.1,III
Merck Index: 12,09273

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR