Tantalum ≥99.95% (metals basis excluding Nb), foil, annealed, Thickness: 0.5 mm (0.02 in)
Supplier: Thermo Scientific Chemicals
10355-FI
10355-GW
10355-GH
10355-GW
10355-FF
10355-GH
10355-FF
10355-FI
CAAA10355-GWEA
3376.46
CAD
CAAA10355-GW
CAAA10355-FF
AA10355-FI
AA10355-GW
AA10355-FF
CAAA10355-GH
AA10355-GH
CAAA10355-FI
Tantalum ≥99.95% (metals basis excluding Nb), foil, annealed, Thickness: 0.5 mm (0.02 in)
Tantalum
Formula:
Ta Boiling Pt: 5427 °C (1013 hPa) Melting Pt: 2996 °C Density: 16.69 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011252 CAS Number: 7440-25-7 Merck Index: 12,09223 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...