Tantalum ≥99.95% (metals basis excluding Nb), wire, annealed, Ø 2.0 mm (0.08 in)
Supplier: Thermo Scientific Chemicals
10354-BU
10354-BY
10354-CC
CAAA10354-BUEA
267.02
CAD
CAAA10354-BU
CAAA10354-BY
CAAA10354-CC
Tantalum ≥99.95% (metals basis excluding Nb), wire, annealed, Ø 2.0 mm (0.08 in)
Tantalum
MDL: MFCD00011252
Purity: 99.95%
CAS No.: 7440-25-7
Physical Form: ≈52.1g/m, Temper: soft
Odor: Odorless
MDL No.: MFCD00011252
Formula:
Ta Boiling Pt: 5427 °C (1013 hPa) Melting Pt: 2996 °C Density: 16.69 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011252 CAS Number: 7440-25-7 EINECS: 231-135-5 Merck Index: 12,09223 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...