Tantalum hard ≥99.95% (metals basis), foil, hard, Thickness: 0.025 mm (0.001 in)

Supplier: Thermo Scientific Chemicals

10351-GJ 10351-IV 10351-IX 010351-FY
CAAA10351-GJEA 221.2 CAD
CAAA10351-GJ CAAA10351-IV CAAA10351-IX AA10351-FY
Tantalum hard ≥99.95% (metals basis), foil, hard, Thickness: 0.025 mm (0.001 in)
Tantalum

MDL: MFCD00011252


Purity: 99.95% CAS No.: 7440-25-7 Physical Form: ≈6.3g/100x150mm, Temper: as rolled Odor: Odorless MDL No.: MFCD00011252

Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427
Melting Pt: 2996
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR