Tantalum 99,95 Metals basis, wire, annealed, Ø 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

10349-G1 10349-G5 10349-G9
CAAA10349-G1EA 77.74 CAD
CAAA10349-G1 CAAA10349-G5 CAAA10349-G9
Tantalum 99,95 Metals basis, wire, annealed, Ø 0.5 mm (0.02 in)
Tantalum

MDL: MFCD00011252


Purity: 99.95% CAS No.: 7440-25-7 Physical Form: ≈3.26g/m Temper: soft Odor: Odorless MDL No.: MFCD00011252

Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427
Melting Pt: 2996
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR